Learn More
Biolegend Purified anti-mouse CD49b
Supplier:  Biolegend 103501
Encompass_Preferred
Purified anti-mouse CD49b [HMα2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IP, Block; Size: 50 ug
Catalog No. 50-162-853