Learn More
Biolegend Purified anti-mouse CD49d
Supplier:  Biolegend 103701
Encompass_Preferred
Purified anti-mouse CD49d [9C10 (MFR4.B)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, Block, IHC-F; Size: 50 ug
Catalog No. 50-162-872