Learn More
Biolegend Purified anti-mouse CD49e
Supplier:  Biolegend 103801
Encompass_Preferred
Purified anti-mouse CD49e [5H10-27(MFR5)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 50 ug
Catalog No. 50-162-879