Learn More
Biolegend Purified anti-mouse CD63
Supplier:  Biolegend 143902
Encompass_Preferred
Purified anti-mouse CD63 [NVG-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC, WB, IHC-F; Size: 100 ug
Catalog No. 50-207-1475