Learn More
Biolegend Purified anti-mouse CD86
Supplier:  Biolegend 105001
Encompass_Preferred
Purified anti-mouse CD86 [GL-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IP, IHC-F, ICC; Size: 50 ug
Catalog No. 50-162-989