Learn More
Biolegend Purified anti-mouse CD8a
Supplier:  Biolegend 100802
Encompass_Preferred
Purified anti-mouse CD8a [5H10-1]; Isotype: Rat (SD) IgG2b, λ; Reactivity: Mouse; Apps: FC, ICC, IHC-F; Size: 500 ug
Catalog No. 50-162-503