Learn More
Biolegend Purified anti-mouse CD8a
Supplier:  Biolegend 100701
Encompass_Preferred
Purified anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF, IHC-F, IP; Size: 50 ug
Catalog No. 50-162-474