Learn More
Biolegend Purified anti-mouse CD90.2
Supplier:  Biolegend 105301
Encompass_Preferred
Purified anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, Depletion, Costim; Size: 50 ug
Catalog No. 50-163-034