Learn More
Biolegend Purified anti-mouse CD90.2 (Thy1.2)
Supplier:  Biolegend 140301
Encompass_Preferred
Purified anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F, ICC, IP; Size: 50 ug
Catalog No. 50-165-119