Learn More
Biolegend Purified anti-mouse CD90
Supplier:  Biolegend 105202
Encompass_Preferred
Purified anti-mouse CD90 [G7]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-163-032