Learn More
Biolegend Purified anti-mouse F4/80
Supplier:  Biolegend 123102
Encompass_Preferred
Purified anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, WB; Size: 500 ug
Catalog No. 50-164-052