Learn More
Biolegend Purified anti-mouse FcεRIα
Supplier:  Biolegend 134302
Encompass_Preferred
Purified anti-mouse FcεRIα [MAR-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-164-727