Learn More
Biolegend Purified anti-mouse H-2Kb
Supplier:  Biolegend 116501
Encompass_Preferred
Purified anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype; Apps: FC, IHC-F, IP; Size: 50 ug
Catalog No. 50-163-662