Learn More
Biolegend Purified anti-mouse H-2Kb/H-2Db
Supplier:  Biolegend 114602
Encompass_Preferred
Purified anti-mouse H-2Kb/H-2Db [28-8-6]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2Dd MHC class I alloantigen; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-163-497