Learn More
Biolegend Purified anti-mouse/human CD11b (Maxpar Ready)
Supplier:  Biolegend 101249
Encompass_Preferred
Purified anti-mouse/human CD11b (Maxpar Ready) [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-206-9552