Learn More
Biolegend Purified anti-mouse/human CD324 (E-Cadherin)
Supplier:  Biolegend 147302
Encompass_Preferred
Purified anti-mouse/human CD324 (E-Cadherin) [DECMA-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Dog, Swine; Apps: FC, ICC, IP, WB, FA; Size: 100 ug
Catalog No. 50-711-916