Learn More
Biolegend Purified anti-mouse I-Ab
Supplier:  Biolegend 116402
Encompass_Preferred
Purified anti-mouse I-Ab [AF6-120.1]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, cross-reacts with H-2k and H-2u haplotypes; Apps: FC, IHC-F, IP; Size: 500 ug
Catalog No. 50-163-645