Learn More
Biolegend Purified anti-mouse IFN-γ
Supplier:  Biolegend 513202
Encompass_Preferred
Purified anti-mouse IFN-γ [H22]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: WB, ELISA, ICC, IP; Size: 100 ug
Catalog No. 50-170-155