Learn More
Biolegend Purified anti-mouse IgE
Supplier:  Biolegend 406902
Encompass_Preferred
Purified anti-mouse IgE [RME-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA, FC; Size: 500 ug
Catalog No. 50-169-177