Learn More
Biolegend Purified anti-mouse IgG2b
Supplier:  Biolegend 406701
Encompass_Preferred
Purified anti-mouse IgG2b [RMG2b-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA, FC; Size: 50 ug
Catalog No. 50-169-171