Learn More
Biolegend Purified anti-mouse IgMa
Supplier:  Biolegend 408602
Encompass_Preferred
Purified anti-mouse IgMa [MA-69]; Isotype: Mouse (C57BL/6) IgG1, κ; Reactivity: Mouse; Apps: FC, IHC, ELISA; Size: 500 ug
Catalog No. 50-169-234