Learn More
Biolegend Purified anti-mouse IL-10
Supplier:  Biolegend 504902
Encompass_Preferred
Purified anti-mouse IL-10 [JES5-2A5]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, WB; Size: 500 ug
Catalog No. 50-169-865