Learn More
Biolegend Purified anti-mouse IL-10
Supplier:  Biolegend 505001
Encompass_Preferred
Purified anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF, IHC; Size: 50 ug
Catalog No. 50-169-870