Learn More
Biolegend Purified anti-mouse IL-12/IL-23 p40
Supplier:  Biolegend 505202
Encompass_Preferred
Purified anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Capture, IHC-F, IP, WB; Size: 500 ug
Catalog No. 50-169-884