Learn More
Biolegend Purified anti-mouse IL-27 p28
Supplier:  Biolegend 516902
Encompass_Preferred
Purified anti-mouse IL-27 p28 [MM27-7B1]; Isotype: Mouse IgG2a; Reactivity: Mouse, Human; Apps: ELISA Capture; Size: 500 ug
Catalog No. 50-208-5901