Learn More
Biolegend Purified anti-mouse IL-4
Supplier:  Biolegend 504102
Encompass_Preferred
Purified anti-mouse IL-4 [11B11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF, IP, IHC, ICC; Size: 500 ug
Catalog No. 50-169-824