Learn More
Biolegend Purified anti-mouse IL-6 (Maxpar Ready)
Supplier:  Biolegend 504509
Encompass_Preferred
Purified anti-mouse IL-6 (Maxpar Ready) [MP5-20F3]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF; Size: 100 ug
Catalog No. 50-208-5648