Learn More
Biolegend Purified anti-mouse IL-6
Supplier:  Biolegend 504502
Encompass_Preferred
Purified anti-mouse IL-6 [MP5-20F3]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF, ELISPOT Capture, Neut, IHC-F; Size: 500 ug
Catalog No. 50-169-850