Learn More
Biolegend Purified anti-mouse LAP (TGF-β1)
Supplier:  Biolegend 141402
Encompass_Preferred
Purified anti-mouse LAP (TGF-β1) [TW7-16B4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, IP, WB; Size: 100 ug
Catalog No. 50-165-199