Learn More
Biolegend Purified anti-mouse Ly-6G/Ly-6C (Gr-1)
Supplier:  Biolegend 108402
Encompass_Preferred
Purified anti-mouse Ly-6G/Ly-6C (Gr-1) [RB6-8C5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC, IP, WB; Size: 500 ug
Catalog No. 50-163-245