Learn More
Biolegend Purified anti-mouse MAIR-V
Supplier:  Biolegend 132702
Encompass_Preferred
Purified anti-mouse MAIR-V [TX70]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-660