Learn More
Biolegend Purified anti-mouse MCP-1
Supplier:  Biolegend 506004
Encompass_Preferred
Purified anti-mouse MCP-1 [4E2/MCP]; Isotype: Hamster IgG; Reactivity: Mouse; Apps: ELISA Capture, WB; Size: 500 ug
Catalog No. 50-208-5731