Learn More
Biolegend Purified anti-mouse Qa-2
Supplier:  Biolegend 121711
Encompass_Preferred
Purified anti-mouse Qa-2 [695H1-9-9]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-207-0468