Learn More
Biolegend Purified anti-mouse/rat CD29
Supplier:  Biolegend 102201
Encompass_Preferred
Purified anti-mouse/rat CD29 [HMβ1-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, IHC-F, IP, Block; Size: 50 ug
Catalog No. 50-162-649