Learn More
Biolegend Purified anti-mouse/rat TNF-α
Supplier:  Biolegend 506101
Encompass_Preferred
Purified anti-mouse/rat TNF-α [TN3-19.12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat, Cross-Reactivity: Rabbit (Lapine); Apps: ELISA Capture, IP, WB; Size: 50 ug
Catalog No. 50-169-946