Learn More
Biolegend Purified anti-mouse TCR Vγ1.1/Cr4
Supplier:  Biolegend 141102
Encompass_Preferred
Purified anti-mouse TCR Vγ1.1/Cr4 [2.11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IP; Size: 100 ug
Catalog No. 50-165-177