Learn More
Biolegend Purified anti-mouse IgM (Maxpar Ready)
Supplier:  Biolegend 406527
Encompass_Preferred
Purified anti-mouse IgM (Maxpar Ready) [RMM-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-207-9764