Learn More
Biolegend Purified anti-Musashi-1
Supplier:  Biolegend 869101
Encompass_Preferred
Purified anti-Musashi-1 [3F8-E6-B9]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P; Size: 25 ug
Catalog No. 50-207-5033