Learn More
Biolegend Purified anti-NALCN
Supplier:  Biolegend 857201
Encompass_Preferred
Purified anti-NALCN [N185/7]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: WB; Size: 25 ug
Catalog No. 50-207-4789