Learn More
Biolegend Purified anti-Neurofilament H (NF-H)
Supplier:  Biolegend 835904
Encompass_Preferred
Purified anti-Neurofilament H (NF-H) [SMI 38]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 100 ug
Catalog No. 50-207-4350