Learn More
Biolegend Purified anti-Neurofilament H (NF-H)
Supplier:  Biolegend 835803
Encompass_Preferred
Purified anti-Neurofilament H (NF-H) [SMI 37]; Isotype: Mouse IgM, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 25 ug
Catalog No. 50-207-4346