Learn More
Biolegend Purified anti-Neurofilament H (NF-H), Phosphorylated
Supplier:  Biolegend 835704
Encompass_Preferred
Purified anti-Neurofilament H (NF-H), Phosphorylated [SMI 36]; Isotype: Mouse IgM, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 25 ug
Catalog No. 50-207-4344