Learn More
Biolegend Purified anti-Neurofilament L (NF-L)
Supplier:  Biolegend 846002
Encompass_Preferred
Purified anti-Neurofilament L (NF-L) [NFL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-207-4489