Learn More
Biolegend Purified anti-Neurogranin, 70-78
Supplier:  Biolegend 848502
Encompass_Preferred
Purified anti-Neurogranin, 70-78 [A16007A]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Rat; Apps: WB, ELISA Capture, IHC-P; Size: 100 ug
Catalog No. 50-207-4543