Learn More
Biolegend Purified anti-NFATC3
Supplier:  Biolegend 690602
Encompass_Preferred
Purified anti-NFATC3 [W15221A]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB; Size: 100 ug
Catalog No. 50-208-7587