Learn More
Biolegend Purified anti-NFIL3
Supplier:  Biolegend 698102
Encompass_Preferred
Purified anti-NFIL3 [W16121C]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB; Size: 100 ug
Catalog No. 50-208-7710