Learn More
Biolegend Purified anti-NLRP2
Supplier:  Biolegend 673902
Encompass_Preferred
Purified anti-NLRP2 [8F10B51]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-208-7312