Learn More
Biolegend Purified anti-Nuclear Pore Complex Proteins
Supplier:  Biolegend 902901
Encompass_Preferred
Purified anti-Nuclear Pore Complex Proteins [MAb414]; Isotype: Mouse IgG1; Reactivity: Vertebrate, Xenopus, Yeast; Apps: ICC, WB, IHC-F, IP, IEM; Size: 100 uL
Catalog No. 50-103-0140