Learn More
Biolegend Purified anti-O-GlcNAc
Supplier:  Biolegend 677902
Encompass_Preferred
Purified anti-O-GlcNAc [RL2]; Isotype: Mouse IgG1, λ; Reactivity: Beta-O-linked N-acetylglucosamine (O-GlcNAc) in all metazoans including plants, animals, and the viruses that infect them; Apps: WB, ICC, IHC, IP; Size: 100 ug
Catalog No. 50-208-7383