Learn More
Biolegend Purified anti-p38 MAPK Phospho (Thr180/Tyr182)
Supplier:  Biolegend 690202
Encompass_Preferred
Purified anti-p38 MAPK Phospho (Thr180/Tyr182) [A16016A]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, ICFC; Size: 100 ug
Catalog No. 50-208-7578